Gene Transcripts

Gene Symbol
C4orf17
Entrez Gene ID
84103
Gene Name
chromosome 4 open reading frame 17
Gene Aliases
Chromosome Location
Chr. 4: 99511004 - 99542303 on Build GRCh38
UniGene ID
Hs.97501
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032149.2 NP_115525.2 2, 3 485
XM_011532315.1 XP_011530617.1 2, 3 465
GenBank mRNA AK093085.1 - 2, 3 477
AK223345.1 - 2, 3 472
AK314711.1 - 2, 3 468
AL136838.1 - 2, 3 446
BC074759.2 - 1, 2 162 30915218
CR533542.1 - 1, 2 123
DB051883.1 - 2, 3 485

View Full Product Details