Gene Transcripts

Gene Symbol
MAML2
Entrez Gene ID
84441
Gene Name
mastermind like transcriptional coactivator 2
Gene Aliases
MAM-3, MAM2, MAM3, MLL-MAML2
Chromosome Location
Chr. 11: 95976593 - 96343245 on Build GRCh38
UniGene ID
Hs.428214
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032427.3 NP_115803.1 5 3898
XM_011543023.2 XP_011541325.1 5 5733
XM_011543024.2 XP_011541326.1 5 2438
GenBank mRNA AB058722.1 - 5 3889
AK308157.1 - 6 3664
AY040322.1 - 5 3889
BC143529.1 - 5 2639 9052038
BC152449.1 - 5 3889 8862580
CR627398.1 - 1 2894

View Full Product Details