Gene Transcripts

Gene Symbol
LINC01547
Entrez Gene ID
84536
Gene Name
long intergenic non-protein coding RNA 1547
Gene Aliases
C21orf67, C21orf69, PRED54
Chromosome Location
Chr. 21: 44933284 - 44939913 on Build GRCh38
UniGene ID
Hs.534504
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027129.1 4 943
GenBank mRNA AF380178.1 - 4 875
AY040088.1 - 5 825
BC128244.1 - 3 657
BC128245.1 - 3 657

View Full Product Details