Gene Transcripts

Gene Symbol
KRBA1
Entrez Gene ID
84626
Gene Name
KRAB-A domain containing 1
Gene Aliases
Chromosome Location
Chr. 7: 149714945 - 149734575 on Build GRCh38
UniGene ID
Hs.299560
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290187.1 NP_001277116.1 10 1367
NM_032534.3 NP_115923.2 10 1666
XM_011516636.2 XP_011514938.1 11 1594
XM_011516637.2 XP_011514939.1 11 1591
XM_011516638.2 XP_011514940.1 11 1550
XM_011516639.2 XP_011514941.1 11 1546
XM_011516640.2 XP_011514942.1 11 1543
XM_011516641.2 XP_011514943.1 10 1482
XM_011516642.2 XP_011514944.1 10 1634
XM_011516643.2 XP_011514945.1 10 1495
GenBank mRNA AB058765.2 - 10 1620
AL831935.1 - 8 1091
BC033229.1 - 3 321
BC041587.1 - 3 321
BC142722.1 - 10 1367 40148879
BC146667.1 - 10 1367 40148146
BC150330.1 - 10 1631 8860497
BC152409.1 - 10 1620 8862512

View Full Product Details