Gene Transcripts

Gene Symbol
SPINK7
Entrez Gene ID
84651
Gene Name
serine peptidase inhibitor, Kazal type 7 (putative)
Gene Aliases
ECG2, ECRG2
Chromosome Location
Chr. 5: 148312427 - 148315919 on Build GRCh38
UniGene ID
Hs.244569
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032566.2 NP_115955.1 4 300
GenBank mRNA AF268198.1 - 4 300
AY359072.1 - 4 287
BC109385.1 - 4 263 40013896
BC110067.1 - 4 263 40013897

View Full Product Details