Gene Transcripts
- Gene Symbol
- MYO18B
- Entrez Gene ID
- 84700
- Gene Name
- myosin XVIIIB
- Gene Aliases
- KFS4
- Chromosome Location
- Chr. 22: 25742133 - 26063847 on Build GRCh38
- UniGene ID
- Hs.417959
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001318245.1 |
NP_001305174.1
|
11, 12 |
2632 |
|
NM_032608.6 |
NP_115997.5
|
11, 12 |
2632 |
|
XM_011530458.2 |
XP_011528760.1
|
12, 13 |
2768 |
|
XM_011530459.2 |
XP_011528761.1
|
12, 13 |
2739 |
|
XM_011530460.2 |
XP_011528762.1
|
11, 12 |
2612 |
|
XM_011530461.2 |
XP_011528763.1
|
12, 13 |
2768 |
|
XM_011530464.2 |
XP_011528766.1
|
12, 13 |
2768 |
|
XM_011530465.2 |
XP_011528767.1
|
11, 12 |
2525 |
|
XM_011530466.2 |
XP_011528768.1
|
12, 13 |
2769 |
|
XM_017029012.1 |
XP_016884501.1
|
12, 13 |
2768 |
|
XM_017029013.1 |
XP_016884502.1
|
12, 13 |
2816 |
|
XM_017029014.1 |
XP_016884503.1
|
12, 13 |
2768 |
|
XM_017029015.1 |
XP_016884504.1
|
11, 12 |
2642 |
|
GenBank mRNA |
AB042648.2 |
- |
7, 8 |
944 |
|
AB075376.1 |
- |
11, 12 |
2632 |
|
AJ310931.2 |
- |
11, 12 |
2623 |
|
AY077700.1 |
- |
11, 12 |
2542 |
|
BC144597.1 |
- |
10, 11 |
2432 |
9053127 |
BC150626.1 |
- |
10, 11 |
2432 |
9056996 |
View Full Product Details