Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001201484.1 | NP_001188413.1 | 13 | 2447 | |
NM_032634.3 | NP_116023.2 | 11 | 3600 | ||
NM_152850.3 | NP_690577.2 | 12 | 1995 | ||
XM_005251619.3 | XP_005251676.1 | 11 | 3225 | ||
XM_017015222.1 | XP_016870711.1 | 12 | 3717 | ||
XM_017015223.1 | XP_016870712.1 | 12 | 2376 | ||
XM_017015224.1 | XP_016870713.1 | 12 | 2640 | ||
GenBank mRNA | AK074064.1 | - | 5 | 4983 | |
AK090433.1 | - | 4 | 6187 | ||
AL833956.1 | - | 11 | 3633 | ||
AW149348.1 | - | 1 | 468 | ||
AY358472.1 | - | 10 | 3206 | ||
BC001030.2 | - | 9 | 1325 | ||
BC013987.2 | - | 10 | 1411 | ||
BC029271.1 | - | 10 | 2871 | ||
BC036916.1 | - | 11 | 1960 | 5203414 | |
BC065282.1 | - | 13 | 2118 | 6172460 |