Gene Transcripts
- Gene Symbol
- LRRCC1
- Entrez Gene ID
- 85444
- Gene Name
- leucine rich repeat and coiled-coil centrosomal protein 1
- Gene Aliases
- CLERC, SAP2, VFL1
- Chromosome Location
- Chr. 8: 85107088 - 85146080 on Build GRCh38
- UniGene ID
- Hs.193115
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_033402.4 |
NP_208325.3
|
4, 5 |
754 |
|
XM_005251318.3 |
XP_005251375.1
|
3, 4 |
543 |
|
XM_017013925.1 |
XP_016869414.1
|
3, 4 |
442 |
|
XM_017013926.1 |
XP_016869415.1
|
3, 4 |
425 |
|
GenBank mRNA |
AB051551.1 |
- |
3, 4 |
1375 |
|
AK056185.1 |
- |
3, 4 |
717 |
|
AK300234.1 |
- |
3, 4 |
549 |
|
AK300874.1 |
- |
3, 4 |
398 |
|
AK302648.1 |
- |
4, 5 |
604 |
|
AK307636.1 |
- |
3, 4 |
388 |
|
BC030701.2 |
- |
4, 5 |
627 |
4837403 |
View Full Product Details