Gene Transcripts
- Gene Symbol
- L3MBTL4
- Entrez Gene ID
- 91133
- Gene Name
- l(3)mbt-like 4 (Drosophila)
- Gene Aliases
- HsT1031
- Chromosome Location
- Chr. 18: 5954703 - 6414911 on Build GRCh38
- UniGene ID
- Hs.128279
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_173464.3 |
NP_775735.2
|
5, 6 |
430 |
|
XM_005258166.1 |
XP_005258223.1
|
5, 6 |
389 |
|
XM_006722364.2 |
XP_006722427.1
|
5, 6 |
389 |
|
XM_011525757.1 |
XP_011524059.1
|
5, 6 |
389 |
|
XM_011525758.1 |
XP_011524060.1
|
5, 6 |
403 |
|
XM_011525759.1 |
XP_011524061.1
|
5, 6 |
410 |
|
XM_011525760.1 |
XP_011524062.1
|
5, 6 |
389 |
|
XM_011525761.1 |
XP_011524063.1
|
5, 6 |
403 |
|
XM_011525762.1 |
XP_011524064.1
|
5, 6 |
389 |
|
XM_011525763.2 |
XP_011524065.1
|
5, 6 |
389 |
|
XM_011525765.2 |
XP_011524067.1
|
5, 6 |
389 |
|
XM_017026074.1 |
XP_016881563.1
|
5, 6 |
389 |
|
XM_017026075.1 |
XP_016881564.1
|
5, 6 |
389 |
|
XM_017026076.1 |
XP_016881565.1
|
5, 6 |
410 |
|
XM_017026077.1 |
XP_016881566.1
|
5, 6 |
389 |
|
GenBank mRNA |
AK093255.1 |
- |
5, 6 |
389 |
|
BC039316.1 |
- |
5, 6 |
422 |
5298402 |
DB455331.1 |
- |
5, 6 |
431 |
|
View Full Product Details