Gene Transcripts
- Gene Symbol
- SHKBP1
- Entrez Gene ID
- 92799
- Gene Name
- SH3KBP1 binding protein 1
- Gene Aliases
- PP203, Sb1
- Chromosome Location
- Chr. 19: 40576851 - 40591399 on Build GRCh38
- UniGene ID
- Hs.26506
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_138392.3 |
NP_612401.2
|
2, 3 |
179 |
|
XM_006723474.1 |
XP_006723537.1
|
2, 3 |
179 |
|
XM_006723475.1 |
XP_006723538.1
|
2, 3 |
179 |
|
XM_011527492.1 |
XP_011525794.1
|
2, 3 |
179 |
|
XM_011527493.1 |
XP_011525795.1
|
1, 2 |
44 |
|
XM_017027473.1 |
XP_016882962.1
|
2, 3 |
179 |
|
XM_017027474.1 |
XP_016882963.1
|
1, 2 |
44 |
|
GenBank mRNA |
AF258553.1 |
- |
2, 3 |
178 |
|
AK075057.1 |
- |
2, 3 |
175 |
|
AK300808.1 |
- |
1, 2 |
117 |
|
AK300819.1 |
- |
2, 3 |
175 |
|
AK312748.1 |
- |
2, 3 |
143 |
|
BC022855.1 |
- |
2, 3 |
150 |
4102901 |
DB038927.1 |
- |
2, 3 |
179 |
|
View Full Product Details