Gene Transcripts
- Gene Symbol
- PKHD1L1
- Entrez Gene ID
- 93035
- Gene Name
- polycystic kidney and hepatic disease 1 (autosomal recessive)-like 1
- Gene Aliases
- PKHDL1
- Chromosome Location
- Chr. 8: 109362477 - 109536809 on Build GRCh38
- UniGene ID
- Hs.170128
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_177531.4 |
NP_803875.2
|
56, 57 |
9572 |
|
XM_011517371.2 |
XP_011515673.1
|
56, 57 |
9523 |
|
XM_017013969.1 |
XP_016869458.1
|
56, 57 |
9523 |
|
XM_017013970.1 |
XP_016869459.1
|
55, 56 |
9409 |
|
XM_017013971.1 |
XP_016869460.1
|
56, 57 |
9361 |
|
XM_017013972.1 |
XP_016869461.1
|
54, 55 |
9220 |
|
XM_017013973.1 |
XP_016869462.1
|
32, 33 |
6579 |
|
XM_017013974.1 |
XP_016869463.1
|
27, 28 |
6079 |
|
GenBank mRNA |
AK131338.1 |
- |
2, 3 |
254 |
|
AY219181.1 |
- |
56, 57 |
9572 |
|
View Full Product Details