Gene Transcripts
- Gene Symbol
- NOSTRIN
- Entrez Gene ID
- 115677
- Gene Name
- nitric oxide synthase trafficking
- Gene Aliases
- DaIP2
- Chromosome Location
- Chr. 2: 168786539 - 168865339 on Build GRCh38
- UniGene ID
- Hs.189780
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001039724.3 |
NP_001034813.2
|
13, 14 |
1246 |
|
NM_001171631.1 |
NP_001165102.1
|
18, 19 |
2118 |
|
NM_001171632.1 |
NP_001165103.1
|
12, 13 |
1162 |
|
NM_052946.3 |
NP_443178.2
|
12, 13 |
1153 |
|
XM_005246270.1 |
XP_005246327.1
|
13, 14 |
1305 |
|
XM_006712233.3 |
XP_006712296.1
|
14, 15 |
1368 |
|
XM_011510540.2 |
XP_011508842.1
|
14, 15 |
1368 |
|
XM_011510543.2 |
XP_011508845.1
|
7, 8 |
821 |
|
XM_017003278.1 |
XP_016858767.1
|
13, 14 |
1197 |
|
XM_017003279.1 |
XP_016858768.1
|
13, 14 |
1691 |
|
XM_017003280.1 |
XP_016858769.1
|
12, 13 |
1168 |
|
GenBank mRNA |
AJ532842.1 |
- |
13, 14 |
1189 |
|
AK002203.2 |
- |
13, 14 |
1218 |
|
AK093444.1 |
- |
18, 19 |
2116 |
|
AK123698.1 |
- |
2, 3 |
1230 |
|
AK290254.1 |
- |
13, 14 |
1212 |
|
BC014189.2 |
- |
12, 13 |
1153 |
4650302 |
BC093072.1 |
- |
13, 14 |
1193 |
30327154 |
BF114987.1 |
- |
3 |
429 |
|
DQ402497.1 |
- |
12, 13 |
1105 |
|
View Full Product Details