Gene Transcripts
- Gene Symbol
- CLNK
- Entrez Gene ID
- 116449
- Gene Name
- cytokine dependent hematopoietic cell linker
- Gene Aliases
- MIST
- Chromosome Location
- Chr. 4: 10489899 - 10734853 on Build GRCh38
- UniGene ID
- Hs.678910
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_052964.2 |
NP_443196.2
|
9, 10 |
602 |
|
XM_011513775.2 |
XP_011512077.1
|
9, 10 |
833 |
|
XM_017007684.1 |
XP_016863173.1
|
9, 10 |
833 |
|
XM_017007685.1 |
XP_016863174.1
|
9, 10 |
833 |
|
GenBank mRNA |
AB032369.1 |
- |
7, 8 |
411 |
|
AB110420.1 |
- |
9, 10 |
705 |
|
AK093001.1 |
- |
9, 10 |
604 |
|
BC029887.1 |
- |
9, 10 |
632 |
|
BC137553.1 |
- |
9, 10 |
531 |
9021553 |
BC137556.1 |
- |
9, 10 |
531 |
9021556 |
View Full Product Details