Gene Transcripts

Gene Symbol
C12orf54
Entrez Gene ID
121273
Gene Name
chromosome 12 open reading frame 54
Gene Aliases
HSD-29, HSD-30
Chromosome Location
Chr. 12: 48413154 - 48496514 on Build GRCh38
UniGene ID
Hs.98202
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152319.3 NP_689532.1 8 464
XM_005268636.3 XP_005268693.1 7 346
XM_011537896.2 XP_011536198.1 10 1112
XM_017018796.1 XP_016874285.1 12 1302
GenBank mRNA AY255793.1 - 7 369
AY255794.1 - 8 568
BC031670.2 - 7 390 5165130
BQ429029.1 - 8 467

View Full Product Details