Gene Transcripts

Gene Symbol
CSNK1A1L
Entrez Gene ID
122011
Gene Name
casein kinase 1 alpha 1 like
Gene Aliases
CK1
Chromosome Location
Chr. 13: 37103260 - 37105664 on Build GRCh38
UniGene ID
Hs.512897
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145203.5 NP_660204.2 1 783
GenBank mRNA BC028723.2 - 1 785

View Full Product Details