Gene Transcripts

Gene Symbol
TPRG1L
Entrez Gene ID
127262
Gene Name
tumor protein p63 regulated 1-like
Gene Aliases
FAM79A, h-mover, mover
Chromosome Location
Chr. 1: 3624992 - 3630131 on Build GRCh38
UniGene ID
Hs.20529
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_182752.3 NP_877429.2 4 682
GenBank mRNA AK025464.1 - 3 402
AK096427.1 - 4 659
AK289919.1 - 4 632
AK298343.1 - 4 521
BC019034.2 - 3 447 4576259

View Full Product Details