Gene Transcripts

Gene Symbol
WFDC12
Entrez Gene ID
128488
Gene Name
WAP four-disulfide core domain 12
Gene Aliases
C20orf122, SWAM2, WAP2, dJ211D12.4
Chromosome Location
Chr. 20: 45123426 - 45124465 on Build GRCh38
UniGene ID
Hs.352180
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080869.1 NP_543145.1 1 79
GenBank mRNA AY037803.1 - 1 79
AY358678.1 - 1 65

View Full Product Details