Gene Transcripts

Gene Symbol
OR13C8
Entrez Gene ID
138802
Gene Name
olfactory receptor family 13 subfamily C member 8
Gene Aliases
OR37H, OR9-10
Chromosome Location
Chr. 9: 104569168 - 104570130 on Build GRCh38
UniGene ID
Hs.553599
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004483.1 NP_001004483.1 1 150

View Full Product Details