Gene Transcripts

Gene Symbol
ODF4
Entrez Gene ID
146852
Gene Name
outer dense fiber of sperm tails 4
Gene Aliases
CT134, CT136, OPPO1
Chromosome Location
Chr. 17: 8339840 - 8346048 on Build GRCh38
UniGene ID
Hs.186045
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001319953.1 NP_001306882.1 2 365
NM_153007.4 NP_694552.2 2 680
XM_005256476.3 XP_005256533.1 3 533
XM_011523688.1 XP_011521990.1 3 302
GenBank mRNA AB081120.1 - 2 680
AY237799.1 - 2 680
BC112010.1 - 2 594 8327478
BC112014.1 - 2 594 8327482
FJ235526.1 - 2 335

View Full Product Details