Gene Transcripts
- Gene Symbol
- MFSD4A
- Entrez Gene ID
- 148808
- Gene Name
- major facilitator superfamily domain containing 4A
- Gene Aliases
- MFSD4, UNQ3064
- Chromosome Location
- Chr. 1: 205568984 - 205602918 on Build GRCh38
- UniGene ID
- Hs.567714
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_181644.4 |
NP_857595.3
|
8, 9 |
1443 |
|
XM_011509229.2 |
XP_011507531.1
|
7, 8 |
1182 |
|
GenBank mRNA |
AK057733.1 |
- |
4, 5 |
513 |
|
AK090831.1 |
- |
2, 3 |
170 |
|
AK091896.1 |
- |
8, 9 |
1384 |
|
AK295148.1 |
- |
7, 8 |
1030 |
|
AK304109.1 |
- |
7, 8 |
1248 |
|
AK304120.1 |
- |
7, 8 |
1109 |
|
AL713676.1 |
- |
6, 7 |
749 |
|
AY358107.1 |
- |
6, 7 |
830 |
|
BC036549.2 |
- |
8, 9 |
1393 |
5264261 |
View Full Product Details