Gene Transcripts

Gene Symbol
C9orf84
Entrez Gene ID
158401
Gene Name
chromosome 9 open reading frame 84
Gene Aliases
Chromosome Location
Chr. 9: 111686175 - 111795949 on Build GRCh38
UniGene ID
Hs.428209
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080551.2 NP_001074020.2 5 888
NM_173521.4 NP_775792.4 7 895
NR_109816.1 9 1097
XM_006716974.2 XP_006717037.1 9 1081
XM_006716975.2 XP_006717038.1 9 1081
XM_011518302.2 XP_011516604.1 7 2890
XM_011518303.1 XP_011516605.1 7 862
XM_011518306.1 XP_011516608.1 4 482
XM_011518309.1 XP_011516611.1 9 1081
XM_017014340.1 XP_016869829.1 7 1699
XM_017014341.1 XP_016869830.1 9 1081
GenBank mRNA AK057341.1 - 9 1032
AK126055.1 - 5 888
AL833535.1 - 7 889
BC112357.1 - 9 1025
BC144680.1 - 9 1021
DB066382.1 - 4 475

View Full Product Details