Gene Transcripts

Gene Symbol
EXD1
Entrez Gene ID
161829
Gene Name
exonuclease 3'-5' domain containing 1
Gene Aliases
EXDL1
Chromosome Location
Chr. 15: 41182728 - 41231074 on Build GRCh38
UniGene ID
Hs.307999
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286441.1 NP_001273370.1 6, 7 768
NM_152596.3 NP_689809.2 4, 5 513
XM_006720415.3 XP_006720478.1 5, 6 685
XM_011521298.2 XP_011519600.1 7, 8 998
XM_011521299.2 XP_011519601.1 7, 8 729
XM_011521300.2 XP_011519602.1 4, 5 257
XM_011521301.2 XP_011519603.1 6, 7 554
XM_017021970.1 XP_016877459.1 4, 5 257
XM_017021971.1 XP_016877460.1 4, 5 255
GenBank mRNA AK292524.1 - 4, 5 419
AK302864.1 - 6, 7 702
BC030628.2 - 4, 5 453 4827307

View Full Product Details