Gene Transcripts

Gene Symbol
ZFP1
Entrez Gene ID
162239
Gene Name
ZFP1 zinc finger protein
Gene Aliases
ZNF475
Chromosome Location
Chr. 16: 75126285 - 75172234 on Build GRCh38
UniGene ID
Hs.388813
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318469.1 NP_001305398.1 4 723
NM_001318471.1 NP_001305400.1 5 779
NM_001318472.1 NP_001305401.1 3 646
NM_001318473.1 NP_001305402.1 3 596
NM_001318474.1 NP_001305403.1 3 577
NM_001318475.1 NP_001305404.1 4 735
NM_001318476.1 NP_001305405.1 4 721
NM_153688.3 NP_710155.2 4 704
XM_011522921.2 XP_011521223.1 6 764
XM_017022983.1 XP_016878472.1 4 891
XM_017022984.1 XP_016878473.1 5 1018
XM_017022985.1 XP_016878474.1 5 1023
XM_017022986.1 XP_016878475.1 6 1111
XM_017022987.1 XP_016878476.1 4 735
GenBank mRNA AK094761.1 - 4 641
AK291372.1 - 3 524
AK296560.1 - 4 653
AK308134.1 - 3 553
AL538214.3 - 4 591
BC033774.1 - 1 67
BC064604.1 - 4 582
BC113863.1 - 2 395
BC114471.1 - 2 395
BC127184.1 - 2 395
BX647144.1 - 2 359
DA781502.1 - 3 500
DC371394.1 - 3 524

View Full Product Details