Gene Transcripts

Gene Symbol
UGT3A2
Entrez Gene ID
167127
Gene Name
UDP glycosyltransferase family 3 member A2
Gene Aliases
Chromosome Location
Chr. 5: 36035017 - 36071358 on Build GRCh38
UniGene ID
Hs.348941
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001168316.1 NP_001161788.1 3 456
NM_174914.3 NP_777574.2 4 558
XM_011513988.2 XP_011512290.1 5 600
XM_017009150.1 XP_016864639.1 4 609
GenBank mRNA AK075383.1 - 4 519
AK300751.1 - 3 417
AY358416.1 - 4 493
BC103924.1 - 4 507 40007344
BC103925.1 - 4 507 40007348

View Full Product Details