Gene Transcripts

Gene Symbol
DDX53
Entrez Gene ID
168400
Gene Name
DEAD-box helicase 53
Gene Aliases
CAGE, CT26
Chromosome Location
Chr. X: 22999961 - 23002089 on Build GRCh38
UniGene ID
Hs.434416
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_182699.3 NP_874358.2 1 1084
GenBank mRNA AY039237.1 - 1 1089
BC051703.1 - 1069 5297066
BC067878.1 - 1 1075 5295819

View Full Product Details