Gene Transcripts

Gene Symbol
SPAG17
Entrez Gene ID
200162
Gene Name
sperm associated antigen 17
Gene Aliases
CT143, PF6
Chromosome Location
Chr. 1: 117953665 - 118185225 on Build GRCh38
UniGene ID
Hs.528821
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_206996.2 NP_996879.1 3 361
XM_006710426.3 XP_006710489.1 3 361
XM_006710427.3 XP_006710490.1 3 361
XM_011540934.1 XP_011539236.1 3 361
XM_011540935.2 XP_011539237.1 1 62
GenBank mRNA AK091816.1 - 3 347
AY555274.1 - 3 347
BX648110.1 - 3 362

View Full Product Details