Gene Transcripts

Gene Symbol
SLC36A1
Entrez Gene ID
206358
Gene Name
solute carrier family 36 member 1
Gene Aliases
Dct1, LYAAT1, PAT1, TRAMD3
Chromosome Location
Chr. 5: 151411344 - 151556318 on Build GRCh38
UniGene ID
Hs.269004
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001308150.1 NP_001295079.1 8 1007
NM_078483.3 NP_510968.2 8 1007
XM_005268386.1 XP_005268443.1 8 977
XM_006714759.3 XP_006714822.1 8 1056
XM_011537580.2 XP_011535882.1 8 1697
XM_011537581.1 XP_011535883.1 9 1125
XM_011537583.2 XP_011535885.1 9 5562
XM_011537584.2 XP_011535886.1 8 1216
XM_011537585.1 XP_011535887.1 9 1072
XM_011537586.2 XP_011535888.1 8 1820
XM_011537587.2 XP_011535889.1 8 1076
XM_011537588.2 XP_011535890.1 8 920
XM_011537589.2 XP_011535891.1 8 1828
XM_011537590.1 XP_011535892.1 8 835
XM_011537591.1 XP_011535893.1 8 810
XM_011537592.2 XP_011535894.1 8 814
XM_011537593.2 XP_011535895.1 9 1070
XM_011537594.1 XP_011535896.1 10 1184
XM_011537595.2 XP_011535897.2 8 1703
XM_017009216.1 XP_016864705.1 8 903
XM_017009217.1 XP_016864706.1 8 1007
XM_017009218.1 XP_016864707.1 7 789
XM_017009219.1 XP_016864708.1 8 1057
GenBank mRNA AF516142.1 - 7 799
AK057340.1 - 8 977
AK309323.1 - 8 979
AY162213.1 - 8 912
AY227111.1 - 9 927
AY227112.1 - 2 295
BC136437.1 - 8 878 9020426
BC136438.1 - 8 878 9020427
BX537963.1 - 8 1004
GU296670.1 - 7 784

View Full Product Details