Gene Transcripts

Gene Symbol
C7orf62
Entrez Gene ID
219557
Gene Name
chromosome 7 open reading frame 62
Gene Aliases
Chromosome Location
Chr. 7: 88794106 - 88797937 on Build GRCh38
UniGene ID
Hs.112877
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152706.3 NP_689919.1 2 652
XM_011515904.2 XP_011514206.1 2 2869
GenBank mRNA BC028365.2 - 2 652

View Full Product Details