Gene Transcripts
- Gene Symbol
- TXNDC8
- Entrez Gene ID
- 255220
- Gene Name
- thioredoxin domain containing 8
- Gene Aliases
- SPTRX-3, TRX6, bA427L11.2
- Chromosome Location
- Chr. 9: 110300900 - 110337887 on Build GRCh38
- UniGene ID
- Hs.147064
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001003936.3 |
NP_001003936.1
|
4, 5 |
353 |
|
NM_001286946.1 |
NP_001273875.1
|
3, 4 |
293 |
|
XM_017014579.1 |
XP_016870068.1
|
4, 5 |
356 |
|
XM_017014581.1 |
XP_016870070.1
|
4, 5 |
356 |
|
XM_017014582.1 |
XP_016870071.1
|
3, 4 |
296 |
|
XM_017014584.1 |
XP_016870073.1
|
4, 5 |
472 |
|
GenBank mRNA |
AF305830.1 |
- |
4, 5 |
293 |
|
BC130549.1 |
- |
4, 5 |
283 |
40146580 |
BC130551.1 |
- |
4, 5 |
283 |
40146582 |
BC144461.1 |
- |
3, 4 |
223 |
9052987 |
View Full Product Details