Gene Transcripts

Gene Symbol
MORN3
Entrez Gene ID
283385
Gene Name
MORN repeat containing 3
Gene Aliases
Chromosome Location
Chr. 12: 121651108 - 121672650 on Build GRCh38
UniGene ID
Hs.434154
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173855.4 NP_776254.3 3 585
XM_005253869.4 XP_005253926.1 4 514
XM_011538213.2 XP_011536515.1 4 525
GenBank mRNA AY177732.1 - 4 496
BC057760.1 - 3 585 30346478

View Full Product Details