Gene Transcripts

Gene Symbol
ZDHHC22
Entrez Gene ID
283576
Gene Name
zinc finger DHHC-type containing 22
Gene Aliases
C14orf59
Chromosome Location
Chr. 14: 77131270 - 77146265 on Build GRCh38
UniGene ID
Hs.525485
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_174976.2 NP_777636.2 2 251
XM_006720119.3 XP_006720182.1 2 481
XM_011536661.2 XP_011534963.1 2 4120
GenBank mRNA AK095612.1 - 2 251
AK294840.1 - 2 250
BC117676.2 - 1 76 40069732
DA496626.1 - 2 251

View Full Product Details