Gene Transcripts
- Gene Symbol
- GSDMA
- Entrez Gene ID
- 284110
- Gene Name
- gasdermin A
- Gene Aliases
- FKSG9, GSDM, GSDM1
- Chromosome Location
- Chr. 17: 39962973 - 39977880 on Build GRCh38
- UniGene ID
- Hs.448873
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_178171.4 |
NP_835465.2
|
5, 6 |
782 |
|
XM_006721832.2 |
XP_006721895.1
|
5, 6 |
735 |
|
XM_011524651.2 |
XP_011522953.1
|
3, 4 |
498 |
|
XM_017024502.1 |
XP_016879991.1
|
5, 6 |
782 |
|
XM_017024503.1 |
XP_016879992.1
|
5, 6 |
782 |
|
XM_017024504.1 |
XP_016879993.1
|
5, 6 |
782 |
|
GenBank mRNA |
AB093591.1 |
- |
4, 5 |
675 |
|
AF307953.1 |
- |
5, 6 |
697 |
|
AK096439.1 |
- |
6, 7 |
784 |
|
BC109197.2 |
- |
6, 7 |
744 |
40005562 |
View Full Product Details