Gene Transcripts

Gene Symbol
OR2Z1
Entrez Gene ID
284383
Gene Name
olfactory receptor family 2 subfamily Z member 1
Gene Aliases
OR19-4, OR2Z2
Chromosome Location
Chr. 19: 8730954 - 8732009 on Build GRCh38
UniGene ID
Hs.553675
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004699.2 NP_001004699.1 1 380
GenBank mRNA BC137046.1 - 1 379
BC151149.1 - 1 380 9021043

View Full Product Details