Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001370.1 | NP_001361.1 | 15 | 2487 | |
XM_006711956.2 | XP_006712019.1 | 15 | 2592 | ||
XM_011532649.2 | XP_011530951.1 | 15 | 2441 | ||
XM_011532650.2 | XP_011530952.1 | 15 | 2442 | ||
XM_011532652.1 | XP_011530954.1 | 15 | 2487 | ||
XM_011532653.1 | XP_011530955.1 | 15 | 2487 | ||
XM_011532654.1 | XP_011530956.1 | 14 | 2220 | ||
XM_011532655.2 | XP_011530957.1 | 11 | 1509 | ||
XM_011532657.1 | XP_011530959.1 | 9 | 1248 | ||
XM_011532660.1 | XP_011530962.1 | 15 | 2487 | ||
XM_017003521.1 | XP_016859010.1 | 15 | 2487 | ||
XM_017003522.1 | XP_016859011.1 | 10 | 2313 | ||
XM_017003523.1 | XP_016859012.1 | 10 | 1480 | ||
XM_017003524.1 | XP_016859013.1 | 15 | 2487 | ||
GenBank mRNA | AK094676.1 | - | 11 | 1700 | |
BC104884.1 | - | 9 | 1393 | ||
BC113424.1 | - | 9 | 1393 | ||
BC143666.1 | - | 9 | 1393 |