Gene Transcripts
- Gene Symbol
- C3orf56
- Entrez Gene ID
- 285311
- Gene Name
- chromosome 3 open reading frame 56
- Gene Aliases
- Chromosome Location
- Chr. 3: 127193131 - 127198185 on Build GRCh38
- UniGene ID
- Hs.591284
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK097460.1 |
- |
2 |
594 |
|
View Full Product Details