Gene Transcripts

Gene Symbol
DPH3
Entrez Gene ID
285381
Gene Name
diphthamide biosynthesis 3
Gene Aliases
DELGIP, DELGIP1, DESR1, DPH3A, KTI11, ZCSL2
Chromosome Location
Chr. 3: 16257061 - 16264989 on Build GRCh38
UniGene ID
Hs.388087
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001047434.2 NP_001040899.1 1 196
NM_206831.2 NP_996662.1 1 196
GenBank mRNA AK022970.1 - 1 176
AK092040.1 - 1 149
BC010181.2 - 1 105 4543414
DA447430.1 - 1 186
DA875039.1 - 1 196

View Full Product Details