Gene Transcripts

Gene Symbol
P2RY8
Entrez Gene ID
286530
Gene Name
purinergic receptor P2Y8
Gene Aliases
P2Y8
Chromosome Location
Chr. X: 1462572 - 1537488 on Build GRCh38
UniGene ID
Hs.111377
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178129.4 NP_835230.1 2 872
XM_005274429.3 XP_005274486.1 2 1306
XM_006724443.3 XP_006724506.2 3 1385
XM_011546178.2 XP_011544480.1 3 989
XM_011546179.2 XP_011544481.1 2 949
GenBank mRNA BC043610.1 - 2 835

View Full Product Details