Gene Transcripts

Gene Symbol
RXFP4
Entrez Gene ID
339403
Gene Name
relaxin/insulin like family peptide receptor 4
Gene Aliases
GPCR142, GPR100, RLN3R2, RXFPR4
Chromosome Location
Chr. 1: 155941689 - 155942834 on Build GRCh38
UniGene ID
Hs.449914
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181885.2 NP_871001.1 1 859
GenBank mRNA AY170824.1 - 1 838
AY288415.1 - 1 838
AY394502.1 - 1 838
BC101507.1 - 1 858 8069013
BC101509.1 - 1 858 8069015
EU432129.1 - 1 838

View Full Product Details