Gene Transcripts

Gene Symbol
LRRC70
Entrez Gene ID
100130733
Gene Name
leucine rich repeat containing 70
Gene Aliases
SLRN
Chromosome Location
Chr. 5: 62578735 - 62581448 on Build GRCh38
UniGene ID
Hs.721348
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181506.4 NP_852607.3 2 557
GenBank mRNA AK123022.1 - 2 557
AY280614.1 - 2 402
AY358733.1 - 2 421
BC067275.1 - 1 86
BC110838.1 - 2 409

View Full Product Details