Gene Transcripts

Gene Symbol
ZBTB41
Entrez Gene ID
360023
Gene Name
zinc finger and BTB domain containing 41
Gene Aliases
FRBZ1, ZNF924
Chromosome Location
Chr. 1: 197153682 - 197200542 on Build GRCh38
UniGene ID
Hs.529439
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_194314.2 NP_919290.2 1 166
NR_135153.1 1 166
XM_011509507.2 XP_011507809.1 1 145
XM_017001205.1 XP_016856694.1 1 145
GenBank mRNA AK128549.1 - 1 145
AY163816.2 - 1 166
BC132818.1 - 1 130 40146840
BC136600.1 - 1 130 9020590

View Full Product Details