Gene Transcripts

Gene Symbol
NHLRC2
Entrez Gene ID
374354
Gene Name
NHL repeat containing 2
Gene Aliases
Chromosome Location
Chr. 10: 113854632 - 113912506 on Build GRCh38
UniGene ID
Hs.369924
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198514.3 NP_940916.2 1, 2 423
XM_011539769.2 XP_011538071.1 1, 2 423
GenBank mRNA AK090631.1 - 1, 2 394
DA821295.1 - 1, 2 424

View Full Product Details