Gene Transcripts
- Gene Symbol
- ARHGEF33
- Entrez Gene ID
- 100271715
- Gene Name
- Rho guanine nucleotide exchange factor 33
- Gene Aliases
- Chromosome Location
- Chr. 2: 38919363 - 38975449 on Build GRCh38
- UniGene ID
- Hs.99841
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
DV080278.1 |
- |
6 |
604 |
|
- Gene Symbol
- MORN2
- Entrez Gene ID
- 729967
- Gene Name
- MORN repeat containing 2
- Gene Aliases
- BLOCK27, MOPT
- Chromosome Location
- Chr. 2: 38875962 - 38882709 on Build GRCh38
- UniGene ID
- Hs.729207
- Species
- Homo sapiens
View Full Product Details