Gene Transcripts

Gene Symbol
LCN15
Entrez Gene ID
389812
Gene Name
lipocalin 15
Gene Aliases
PRO6093, UNQ2541
Chromosome Location
Chr. 9: 136759634 - 136764513 on Build GRCh38
UniGene ID
Hs.413902
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_203347.1 NP_976222.1 5, 6 552
XM_011518672.1 XP_011516974.1 5, 6 480
GenBank mRNA AY358618.1 - 5, 6 552
BC035124.1 - 4, 5 2681 5263500
BC093893.1 - 5, 6 547 7939738
BC093895.1 - 5, 6 547 7939740

View Full Product Details