Gene Transcripts

Gene Symbol
OR52A5
Entrez Gene ID
390054
Gene Name
olfactory receptor family 52 subfamily A member 5
Gene Aliases
OR11-33
Chromosome Location
Chr. 11: 5131692 - 5132642 on Build GRCh38
UniGene ID
Hs.546593
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005160.2 NP_001005160.1 1 632
GenBank mRNA AY726590.1 - 1 568

View Full Product Details