Gene Transcripts

Gene Symbol
C12orf76
Entrez Gene ID
400073
Gene Name
chromosome 12 open reading frame 76
Gene Aliases
Chromosome Location
Chr. 12: 110041177 - 110067695 on Build GRCh38
UniGene ID
Hs.44817
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207435.2 NP_997318.1 3 535
XM_017019297.1 XP_016874786.1 2 203
GenBank mRNA AK097461.1 - 3 535
BC133010.1 - 2 318 40147032
BC133012.1 - 2 318 40147034

View Full Product Details