Gene Transcripts
- Gene Symbol
- BLID
- Entrez Gene ID
- 414899
- Gene Name
- BH3-like motif containing, cell death inducer
- Gene Aliases
- BRCC2
- Chromosome Location
- Chr. 11: 122115354 - 122116215 on Build GRCh38
- UniGene ID
- Hs.686109
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF303179.1 |
- |
1 |
333 |
|
View Full Product Details