Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004045.3 | NP_004036.1 | 2 | 157 | |
GenBank mRNA | AK128210.1 | - | 2 | 1034 | |
AK293063.1 | - | 2 | 164 | ||
BC112248.1 | - | 2 | 239 | 8327716 | |
BC112250.1 | - | 2 | 239 | 8327718 | |
BI599233.1 | - | 2 | 165 | ||
BT009786.1 | - | 1 | 59 | ||
BU172675.1 | - | 2 | 535 | ||
CN280769.1 | - | 2 | 134 | ||
CR456901.1 | - | 1 | 59 |