Gene Transcripts

Gene Symbol
CALCB
Entrez Gene ID
797
Gene Name
calcitonin related polypeptide beta
Gene Aliases
CALC2, CGRP-II, CGRP2
Chromosome Location
Chr. 11: 15073600 - 15078631 on Build GRCh38
UniGene ID
Hs.534305
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000728.3 NP_000719.1 3 188
GenBank mRNA AK054728.1 - 8 1410
AK291188.1 - 3 191
BC092468.1 - 3 192 5313089
BI668461.1 - 3 197

View Full Product Details