Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001234.4 | NP_001225.1 | 1 | 172 | |
NM_033337.2 | NP_203123.1 | 1 | 172 | ||
GenBank mRNA | AF036365.1 | - | 1 | 142 | |
AF043101.1 | - | 1 | 92 | ||
BC102033.1 | - | 1 | 132 | 40033505 | |
BC102036.1 | - | 1 | 132 | 40033114 | |
BC102037.1 | - | 1 | 132 | 40033118 | |
Y14747.1 | - | 1 | 106 |