Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001804.2 | NP_001795.2 | 2 | 582 | |
GenBank mRNA | AF239666.1 | - | 2 | 567 | |
BC096251.1 | - | 3 | 199 | ||
BC096252.1 | - | 3 | 245 | 40005955 | |
BC127122.1 | - | 3 | 245 | 40005957 | |
U15212.1 | - | 2 | 300 | ||
U51095.1 | - | 2 | 567 |